Tisk…

Celastrol (from Tripterygium wilfordii Hook.F)

Dodavatel: Apollo Scientific

BIC1002-250MG BIC1002-500MG BIC1002-1G
APOSBIC1002-250MGEA 3740 CZK
APOSBIC1002-250MG APOSBIC1002-500MG APOSBIC1002-1G
Celastrol (from Tripterygium wilfordii Hook.F)
Celastrol
Vzorec: C₂₉H₃₈O₄
Storage Temperature: Freezer
MDL Number: MFCD03424073
CAS číslo: 34157-83-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR