Tisk…

Folinic acid calcium salt ≥97.5%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Formyl-5,6,7,8-tetrahydrofolic acid calcium salt, Calcium (4-(((2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl)amino)benzoyl)-L-glutamate, Folinic acid calcium, Calcium folinate

BIC0607-1G BIC0607-5G BIC0607-25G
APOSBIC0607-1GEA 1290 CZK
APOSBIC0607-1G APOSBIC0607-5G APOSBIC0607-25G
Folinic acid calcium salt ≥97.5%
Folinic acid calcium salt
Vzorec: C₂₀H₂₁CaN₇O₇
Storage Temperature: Ambient
MDL Number: MFCD00006704
CAS číslo: 1492-18-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR