Tisk…

Chaetocin

Dodavatel: Apollo Scientific
Varování

Synonyms: (3S,3′S,5aR,5′aR,10bR,10′bR,11aS,11′aS)-2,2′,3,3′,5a,5′a,6,6′-Octahydro-3,3′-bis(hydroxymethyl)-2,2′-dimethyl[10b,10′b(11H,11′H)-bi-3,11a-epidithio-11aH-pyrazino[1′,2′:1,5]pyrrolo[2,3-b]indole]-1,1′,4,4′-tetrone

BIC0128-1MG BIC0128-5MG
APOSBIC0128-1MGEA 2030 CZK
APOSBIC0128-1MG APOSBIC0128-5MG
Chaetocin
Chaetocin

Source: From Chaetomium sp.

Vzorec: C₃₀H₂₈N₆O₆S₄
MW: 696,839 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00133163
CAS číslo: 28097-03-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR