Tisk…

Cefsulodin sodium salt

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Cefsulodin sodium, Sodium (6R,7R)-3-((4-carbamoylpyridin-1-ium-1-yl)methyl)-8-oxo-7-((R)-2-phenyl-2-sulphonatoacetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Sulcephalosporin

BIC0112-1G BIC0112-500MG BIC0112-250MG
APOSBIC0112-1GEA 4960 CZK
APOSBIC0112-1G APOSBIC0112-500MG APOSBIC0112-250MG
Cefsulodin sodium salt
Cefsulodin sodium salt
Vzorec: C₂₂H₁₉N₄NaO₈S₂
Bod tání: 175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07793338
CAS číslo: 52152-93-9
EINECS: 257-692-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR