Tisk…

tert-Butyl-2-aminoethyl(ethyl)carbamate hydrochloride ≥99%

Dodavatel: Apollo Scientific

Synonyms: Boc,Et-EDA.HCl

BIBP1037-1G BIBP1037-5G
APOSBIBP1037-1GEA 3380 CZK
APOSBIBP1037-1G APOSBIBP1037-5G
tert-Butyl-2-aminoethyl(ethyl)carbamate hydrochloride ≥99%
tert-Butyl-2-aminoethyl(ethyl)carbamate hydrochloride
Vzorec: C₉H₂₁ClN₂O₂
MW: 224,73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06796900
CAS číslo: 1001020-32-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR