Tisk…

t-Boc-N-amido-PEG3-amine ≥98%

Dodavatel: Apollo Scientific

Synonyms: Boc-1-amino-4,7,10-trioxa-13-tridecanamine, tert-Butyl (3-(2-(2-(3-aminopropoxy)ethoxy)ethoxy)propyl)carbamate, N-Boc-C1-PEG3-C3-NH2

BIBP1032-5G
APOSBIBP1032-5GEA 4200 CZK
APOSBIBP1032-5G
t-Boc-N-amido-PEG3-amine ≥98%
t-Boc-N-amido-PEG3-amine
Vzorec: C₁₅H₃₂N₂O₅
MW: 320,43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973133
CAS číslo: 194920-62-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR