Tisk…

Boc-8-amino-3,6-dioxaoctanoic acid DCHA ≥98%

Dodavatel: Apollo Scientific

Synonyms: Boc-AEEA.DCHA, Dicyclohexylamine 2,2-dimethyl-4-oxo-3,8,11-trioxa-5-azatridecan-13-oate

BIBP1014-1G BIBP1014-5G
APOSBIBP1014-1GEA 3710 CZK
APOSBIBP1014-1G APOSBIBP1014-5G
Boc-8-amino-3,6-dioxaoctanoic acid DCHA ≥98%
Boc-8-amino-3,6-dioxaoctanoic acid DCHA
Vzorec: C₂₃H₄₄N₂O₆
MW: 444.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06796877
CAS číslo: 560088-79-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR