(2S,3R,4S,5S)-2-(4-Nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol
Dodavatel: Apollo Scientific
BIB6309-250MG
BIB6309-500MG
BIB6309-1G
APOSBIB6309-250MGEA
3940
CZK
APOSBIB6309-250MG
APOSBIB6309-500MG
APOSBIB6309-1G
(2S,3R,4S,5S)-2-(4-Nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol
(2S,3R,4S,5S)-2-(4-Nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol
Vzorec:
C₁₁H₁₃NO₇ Bod tání: 201…202 °C Storage Temperature: Freezer |
MDL Number:
MFCD00151486 CAS číslo: 1223-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...