Tisk…

N-Ethyl-N-methylcarbamoyl chloride

Dodavatel: Apollo Scientific
Varování

Synonyms: N-Ethyl-N-methylcarbamoyl Chloride

BIB6299-100G BIB6299-25G BIB6299-50G
APOSBIB6299-100GEA 13230 CZK
APOSBIB6299-100G APOSBIB6299-25G APOSBIB6299-50G
N-Ethyl-N-methylcarbamoyl chloride
N-Ethyl-N-methylcarbamoyl chloride
Vzorec: C₄H₈ClNO
MW: 121,56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061404
CAS číslo: 42252-34-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR