Tisk…

6-TAMRA

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-4-carboxybenzoate

BIB6288-250MG BIB6288-50MG BIB6288-100MG
APOSBIB6288-250MGEA 7250 CZK
APOSBIB6288-250MG APOSBIB6288-50MG APOSBIB6288-100MG
6-TAMRA
6-TAMRA
Vzorec: C₂₅H₂₂N₂O₅
MW: 430,45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01073638
CAS číslo: 91809-67-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR