Tisk…

5-Carboxytetramethylrhodamine (5-TAMRA)

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-TAMRA, 5-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-5-carboxybenzoate, 2-[3,6-Bis(dimethylamino)xanthen-10-ium-9-yl]-5-carboxybenzoate, TAMRA, 5-isomer

BIB6287-100MG BIB6287-1G BIB6287-500MG
APOSBIB6287-100MGEA 2070 CZK
APOSBIB6287-100MG APOSBIB6287-1G APOSBIB6287-500MG
5-Carboxytetramethylrhodamine (5-TAMRA)
5-Carboxytetramethylrhodamine (5-TAMRA)
Vzorec: C₂₅H₂₂N₂O₅
MW: 430,45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00269769
CAS číslo: 91809-66-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR