Tisk…

8-Acetoxypyrene-1,3,6-trisulphonic acid trisodium salt

Dodavatel: Apollo Scientific
Varování

BIB6286-10G BIB6286-1G BIB6286-5G
APOSBIB6286-10GEA 32870 CZK
APOSBIB6286-10G APOSBIB6286-1G APOSBIB6286-5G
8-Acetoxypyrene-1,3,6-trisulphonic acid trisodium salt
8-Acetoxypyrene-1,3,6-trisulphonic acid trisodium salt
Vzorec: C₁₈H₉Na₃O₁₁S₃
MW: 566.42 g/mol
Storage Temperature: Freezer
CAS číslo: 115787-83-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR