AQC
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Aminoquinoline N-succinimidyl ester, 2,5-Dioxo-1-pyrrolidinyl 6-Quinolylcarbamate
BIB6284-100MG
BIB6284-500MG
BIB6284-1G
APOSBIB6284-100MGEA
7870
CZK
APOSBIB6284-100MG
APOSBIB6284-500MG
APOSBIB6284-1G
AQC
AQC
Vzorec:
C₁₄H₁₁N₃O₄ MW: 285.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01940654 CAS číslo: 148757-94-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...