Tisk…

5-Methyl-DL-tryptophan

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Amino-3-(5-methyl-1H-indol-3-yl)propanoic acid, H-DL-Trp(5-Me)-OH

BIB6112-1G BIB6112-500MG BIB6112-5G
APOSBIB6112-1GEA 7440 CZK
APOSBIB6112-1G APOSBIB6112-500MG APOSBIB6112-5G
5-Methyl-DL-tryptophan
5-Methyl-DL-tryptophan
Vzorec: C₁₂H₁₄N₂O₂
MW: 218,26 g/mol
Bod tání: 280…282 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005652
CAS číslo: 951-55-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR