L(-)-Malic acid disodium salt
Dodavatel: Apollo Scientific
|
Varování
|
BIB6095-100G
BIB6095-25G
APOSBIB6095-100GEA
4960
CZK
APOSBIB6095-100G
APOSBIB6095-25G
L(-)-Malic acid disodium salt
L(-)-Malic acid disodium salt
Vzorec:
C₄H₄Na₂O₅ Storage Temperature: Ambient |
CAS číslo:
138-09-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...