2-Keto-D-gluconic acid hemicalcium salt monohydrate
Dodavatel: Apollo Scientific
Synonyms:
Calcium 2-keto-D-gluconate, D-Arabino-2-hexulopyranosonic acid hemicalcium Salt
BIB6086-10G
BIB6086-1G
BIB6086-5G
APOSBIB6086-10GEA
16960
CZK
APOSBIB6086-10G
APOSBIB6086-1G
APOSBIB6086-5G
2-Keto-D-gluconic acid hemicalcium salt monohydrate
2-Keto-D-gluconic acid hemicalcium salt monohydrate
Vzorec:
C₆H₉O₇·½Ca·H₂O MW: 231.22 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00149104 CAS číslo: 3470-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...