Tisk…

myo-Inozitol

Dodavatel: Apollo Scientific

Synonyms: i-Inositol, D-Myo-Inositol, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol, Inositol

BIB6083-100G BIB6083-1KG BIB6083-500G
APOSBIB6083-100GEA 1040 CZK
APOSBIB6083-100G APOSBIB6083-1KG APOSBIB6083-500G
myo-Inozitol
myo-Inozitol
Vzorec: C₆H₁₂O₆
MW: 180,16 g/mol
Bod tání: 222…227 °C
Hustota: 0,63 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00077932
CAS číslo: 87-89-8
EINECS: 201-781-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR