Hematin (from porcine)
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Hydroxyhemin, Ferrihemate
BIB6063-1G
BIB6063-5G
APOSBIB6063-1GEA
2480
CZK
APOSBIB6063-1G
APOSBIB6063-5G
Hematin (from porcine)
Hematin
Vzorec:
C₃₄H₃₃FeN₄O₅ Storage Temperature: Freezer |
MDL Number:
MFCD00011615 CAS číslo: 15489-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...