Tisk…

Sodium glycocholate

Dodavatel: Apollo Scientific

Synonyms: Glycocholic acid, sodium salt hydrate, Sodium ((4R)-4-((3R,7R,10S,12S,13R,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoyl)glycinate, Glycocholic acid sodium

BIB6061-10G BIB6061-5G
APOSBIB6061-10GEA 2200 CZK
APOSBIB6061-10G APOSBIB6061-5G
Sodium glycocholate
Sodium glycocholate

Bile salt.

Vzorec: C₂₆H₄₂NNaO₆
Bod tání: 210…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00036741
CAS číslo: 863-57-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR