Tisk…

DL-Glyceraldehyde dimer 93+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3,6-Dihydroxy-1,4-dioxane-2,5-dimethanol, (2R,3S,5S,6R)-3,6-Bis(hydroxymethyl)-1,4-dioxane-2,5-diol

BIB6059-10G BIB6059-1G BIB6059-5G
APOSBIB6059-10GEA 28770 CZK
APOSBIB6059-10G APOSBIB6059-1G APOSBIB6059-5G
DL-Glyceraldehyde dimer 93+%
DL-Glyceraldehyde dimer
Vzorec: C₆H₁₂O₆
MW: 180,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070528
CAS číslo: 23147-59-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR