Tisk…

D-(+)-Fucose

Dodavatel: Apollo Scientific

Synonyms: alpha-D-Fucose

BIB6052-1G
APOSBIB6052-1GEA 3060 CZK
APOSBIB6052-1G
D-(+)-Fucose
D-(+)-Fucose
Vzorec: C₆H₁₂O₅
Bod tání: 144…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00135603
CAS číslo: 3615-37-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR