Tisk…

1,2-Dipalmitoyl-sn-glycero-3-phosphocholine

Dodavatel: Apollo Scientific

Synonyms: 2,3-Bis(palmitoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, (R)-2,3-Bis(palmitoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate, L-DPPC, 1,2-Dihexadecanoyl-sn-glycero-3-phosphocholine, L-α-Lecithin, Dipalmitoyl, L-alpha-Dipalmitoyl phosphatidylcholine

BIB6043-1G BIB6043-250MG BIB6043-10G
APOSBIB6043-1GEA 6850 CZK
APOSBIB6043-1G APOSBIB6043-250MG APOSBIB6043-10G
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine
Vzorec: C₄₀H₈₀NO₈P
MW: 734.05 g/mol
Bod tání: 220 °C
Storage Temperature: Freezer
MDL Number: MFCD00036903
CAS číslo: 63-89-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR