Tisk…

L-Dehydroascorbic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (5R)-5-[(1S)-1,2-dihydroxyethyl]oxolane-2,3,4-trione, L-Dehydroascorbate, L-Threo-hexo-2,3-diulosono-1,4-lactone

BIB6039-1G BIB6039-5G BIB6039-10G
APOSBIB6039-1GEA 2480 CZK
APOSBIB6039-1G APOSBIB6039-5G APOSBIB6039-10G
L-Dehydroascorbic acid
L-Dehydroascorbic acid
Vzorec: C₆H₆O₆
MW: 174,108 g/mol
Bod tání: 228 °C
Storage Temperature: Freezer
MDL Number: MFCD00066467
CAS číslo: 490-83-5
EINECS: 207-720-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR