Tisk…

Chlorogenic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (1S,3R,4R,5R)-3-(((3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1,4,5-trihydroxycyclohexanecarboxylic acid, 3-O-Caffeoylquinic acid, 3-(3,4-Dihydroxycinnamoyl)quinic acid, (1S,3R,4R,5R)-3-[[3-(3,4-Dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4,5-trihydroxycyclohexanecarboxylic acid

BIB6027-25G BIB6027-1G BIB6027-5G
APOSBIB6027-25GEA 4400 CZK
APOSBIB6027-25G APOSBIB6027-1G APOSBIB6027-5G
Chlorogenic acid
Chlorogenic acid
Vzorec: C₁₆H₁₈O₉
MW: 354,31 g/mol
Bod tání: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00003862
CAS číslo: 327-97-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR