Tisk…

Thio-NAD 92+%

Dodavatel: Apollo Scientific
Varování

Synonyms: (2R,3R,4S,5R)-5-[[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxymethyl]-2-(3-carbamothioylpyridin-1-ium-1-yl)-4-hydroxyoxolan-3-olate, Thionicotinamide-DPN, Thionicotinamide Adenine dinucleotide oxidized form

BIB5004-100MG BIB5004-25MG
APOSBIB5004-100MGEA 15210 CZK
APOSBIB5004-100MG APOSBIB5004-25MG
Thio-NAD 92+%
Thio-NAD

Purity determined enzymatic method (ADH).

Vzorec: C₂₁H₂₇N₇O₁₃P₂S
Storage Temperature: Freezer
MDL Number: MFCD00079609
CAS číslo: 4090-29-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR