Tisk…

Brefeldin A

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural

BIB3042-25MG BIB3042-50MG
APOSBIB3042-25MGEA 2900 CZK
APOSBIB3042-25MG APOSBIB3042-50MG
Brefeldin A
Brefeldin A

BFA is a macrolide antiobiotic. Disrupts mechanism of Golgi apparatus.

Vzorec: C₁₆H₂₄O₄
Bod tání: 200…205 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00083258
CAS číslo: 20350-15-6
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR