MOPSO (3-(4-morpholino)-2-hydroxypropanesulphonic acid) ≥99.5%, Ultrapure
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Hydroxy-3-morpholinopropane-1-sulphonic acid
BIB2401-100G
BIB2401-25G
BIB2401-500G
APOSBIB2401-100GEA
705
CZK
APOSBIB2401-100G
APOSBIB2401-25G
APOSBIB2401-500G
MOPSO (3-(4-morpholino)-2-hydroxypropanesulphonic acid) ≥99.5%, Ultrapure
MOPSO (3-(4-morpholino)-2-hydroxypropanesulphonic acid)
Biological buffer - useful range 6.2 - 7.6
U.S. Patent No 4,246,194 (To Research Organics)
Vzorec:
C₇H₁₅NO₅S MW: 225,27 g/mol Bod tání: 276…279 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009629 CAS číslo: 68399-77-9 EINECS: 269-989-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...