(4-Nitrophenyl)-α-D-galactopyranoside
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol, 4-Nitrophenyl α-D-galactopyranoside
BIB1433-1G
BIB1433-500MG
BIB1433-5G
APOSBIB1433-1GEA
2690
CZK
APOSBIB1433-1G
APOSBIB1433-500MG
APOSBIB1433-5G
(4-Nitrophenyl)-α-D-galactopyranoside
(4-Nitrophenyl)-α-D-galactopyranoside
Vzorec:
C₁₂H₁₅NO₈ MW: 301,26 g/mol Bod tání: 166 °C Storage Temperature: Freezer |
MDL Number:
MFCD00065050 CAS číslo: 7493-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...