Tisk…

2-Naphthyl-ß-D-galactopyranoside

Dodavatel: Apollo Scientific
Varování

BIB1427-250MG BIB1427-500MG
APOSBIB1427-250MGEA 2980 CZK
APOSBIB1427-250MG APOSBIB1427-500MG
2-Naphthyl-ß-D-galactopyranoside
2-Naphthyl-ß-D-galactopyranoside
Vzorec: C₁₆H₁₈O₆
Storage Temperature: Freezer
MDL Number: MFCD00004056
CAS číslo: 33993-25-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR