Naphthol AS-D chloroacetate
Dodavatel: Apollo Scientific
Synonyms:
3-[[(2-Methylphenyl)amino]carbonyl]-2-naphthyl chloroacetate
BIB1424-1G
BIB1424-500MG
APOSBIB1424-1GEA
4570
CZK
APOSBIB1424-1G
APOSBIB1424-500MG
Naphthol AS-D chloroacetate
Naphthol AS-D chloroacetate
Vzorec:
C₂₀H₁₆ClNO₃ MW: 353,8 g/mol Bod tání: 163…171 °C Storage Temperature: Freezer |
CAS číslo:
35245-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...