Tisk…

Sodium-2-bromoethanesulphonate

Dodavatel: Apollo Scientific
Varování

BIB1184-100G BIB1184-1KG BIB1184-500G
APOSBIB1184-100GEA 1240 CZK
APOSBIB1184-100G APOSBIB1184-1KG APOSBIB1184-500G
Sodium-2-bromoethanesulphonate
Sodium-2-bromoethanesulphonate
Vzorec: C₂H₅BrO₃S
Storage Temperature: Ambient
MDL Number: MFCD00007530
CAS číslo: 4263-52-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR