Tisk…

4-Bromo-A-23187

Dodavatel: Apollo Scientific

Synonyms: Antibiotic A-23187, 4-bromo

BIB1001-1MG BIB1001-5MG
APOSBIB1001-1MGEA 4100 CZK
APOSBIB1001-1MG APOSBIB1001-5MG
4-Bromo-A-23187
4-Bromo-A-23187

Calcium ionophore, analog of calcium ionophore A23187.

Vzorec: C₂₉H₃₆BrN₃O₆
MW: 602,52 g/mol
Storage Temperature: Refrigerator
CAS číslo: 76455-48-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR