Tisk…

Artemisinin (from Artemisia annua)

Dodavatel: Apollo Scientific
Varování

BIA4320-100MG BIA4320-250MG BIA4320-500MG
APOSBIA4320-100MGEA 5200 CZK
APOSBIA4320-100MG APOSBIA4320-250MG APOSBIA4320-500MG
Artemisinin (from Artemisia annua)
Artemisinin
Vzorec: C₁₅H₂₂O₅
Bod tání: 156…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00081057
CAS číslo: 63968-64-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR