Tisk…

Ascomycin

Dodavatel: Apollo Scientific
Varování

Synonyms: (3S,4R,5S,8R,9E,12S,14S,15R,16S,18R,19R,26aS)-8-Ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-3-[(1E)-2-[(1R,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethenyl]-14,16-dimethoxy-4,10,12,18-tetramethyl-15,19-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21(4H,23H)-tetrone

BIA4317-50MG
APOSBIA4317-50MGEA 1700 CZK
APOSBIA4317-50MG
Ascomycin
Ascomycin
Vzorec: C₄₃H₆₉NO₁₂
MW: 790 g/mol
Storage Temperature: Freezer
MDL Number: MFCD06198665
CAS číslo: 104987-12-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR