Tisk…

H-Arg-AMC·2HCl

Dodavatel: Apollo Scientific

Synonyms: L-Arginine 7-amido-4-methylcoumarin dihydrochloride

BIA4020-1G BIA4020-500MG
APOSBIA4020-1GEA 10350 CZK
APOSBIA4020-1G APOSBIA4020-500MG
H-Arg-AMC·2HCl
H-Arg-AMC·2HCl
Vzorec: C₁₆H₂₁N₅O₃·2HCl
MW: 404,3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00155578
CAS číslo: 113712-08-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR