4-Aminophenylphosphate sodium salt
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-APP
BIA4012-100MG
BIA4012-1G
BIA4012-500MG
APOSBIA4012-100MGEA
2070
CZK
APOSBIA4012-100MG
APOSBIA4012-1G
APOSBIA4012-500MG
4-Aminophenylphosphate sodium salt
4-Aminophenylphosphate sodium salt
Vzorec:
C₆H₇NO₄P·Na·4.0H₂O Bod tání: 176 °C Storage Temperature: Freezer |
CAS číslo:
108084-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...