Tisk…

2-Aminoisobutyric acid

Dodavatel: Apollo Scientific
Varování

Synonyms: α-Aminoisobutyric acid, 2-Amino-2-methylpropionic acid, 2-Metilalanina, H-Aib-OH, 2-Methylalanine, 2-Amino-2-methylpropanoic acid

BIA4009-100G BIA4009-1KG BIA4009-250G
APOSBIA4009-100GEA 744 CZK
APOSBIA4009-100G APOSBIA4009-1KG APOSBIA4009-250G
2-Aminoisobutyric acid
2-Aminoisobutyric acid
Vzorec: C₄H₉NO₂
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00008049
CAS číslo: 62-57-7
EINECS: 200-544-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR