Tisk…

Adenosine 99%

Dodavatel: Apollo Scientific
Varování

BIA2545-100G BIA2545-10G BIA2545-25G
APOSBIA2545-100GEA 2480 CZK
APOSBIA2545-100G APOSBIA2545-10G APOSBIA2545-25G
Adenosine 99%
Adenosine

Endogenous neurotransmitter.

Vzorec: C₁₀H₁₃N₅O₄
Bod tání: 234…236 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005752
CAS číslo: 58-61-7
EINECS: 200-389-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR