Tisk…

Adenin ≥99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine

BIA2543-100G BIA2543-25G BIA2543-500G
APOSBIA2543-100GEA 1240 CZK
APOSBIA2543-100G APOSBIA2543-25G APOSBIA2543-500G
Adenin ≥99%
Adenin

Cytokin growth regulator.


  • Plant tissue culture tested

Vzorec: C₅H₅N₅
MW: 135,13 g/mol
Bod tání: 360…365 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00041790
CAS číslo: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR