8-Aminonaphthalene-1,3,6-trisulphonic acid disodium salt 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
8-Amino-1,3,6-naphthalenetrisulphonic acid disodium salt, Koch acid disodium salt, Amino H acid disodium salt, 1-Naphthylamine-3,6,8-trisulphonic acid disodium salt, Disodium 8-Amino-1,3,6-naphthalenetrisulphonate, ANTS, Disodium-8-amino-1,3,6-naphthalenetrisulphonate
BIA1386-10G
BIA1386-1G
BIA1386-5G
APOSBIA1386-10GEA
3640
CZK
APOSBIA1386-10G
APOSBIA1386-1G
APOSBIA1386-5G
8-Aminonaphthalene-1,3,6-trisulphonic acid disodium salt 98%
8-Aminonaphthalene-1,3,6-trisulphonic acid disodium salt
Fluorescent label for glycoproteins andsaccharides.
Vzorec:
C₁₀H₉NO₉S₃ MW: 427,32 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00070572 CAS číslo: 5398-34-5 EINECS: 226-431-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...