Tisk…

(S)-2-(4-Aminobenzamido)pentanedioic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: N-(4-Aminobenzoyl)-L-glutamic acid, N-(4-NH₂-Bz)-Glu-OH

BIA1350-100G BIA1350-25G
APOSBIA1350-100GEA 705 CZK
APOSBIA1350-100G APOSBIA1350-25G
(S)-2-(4-Aminobenzamido)pentanedioic acid
(S)-2-(4-Aminobenzamido)pentanedioic acid
Vzorec: C₁₂H₁₄N₂O₅
MW: 266.2500 g/mol
Bod tání: 175 °C
Storage Temperature: Freezer
MDL Number: MFCD00042821
CAS číslo: 4271-30-1
EINECS: 224-261-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR