Acetyl-L-carnitine hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(R)-3-Acetoxy-4-(trimethylammonio)butyrate Hydrochloride, (R)-2-Acetoxy-3-carboxy-N,N,N-trimethylpropan-1-aminium chloride, Acetyl-L-carnitine HCl
BIA1225-100G
BIA1225-250G
BIA1225-25G
APOSBIA1225-100GEA
4570
CZK
APOSBIA1225-100G
APOSBIA1225-250G
APOSBIA1225-25G
Acetyl-L-carnitine hydrochloride
Acetyl-L-carnitine hydrochloride
Vzorec:
C₉H₁₇NO₄ Bod tání: 194 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00082230 CAS číslo: 5080-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...