Tisk…

N-Acetyl-D-alanine

Dodavatel: Apollo Scientific
Varování

Synonyms: N-Ac-D-Ala-OH, Ac-D-Ala-OH

BIA1217-1G BIA1217-5G
APOSBIA1217-1GEA 622 CZK
APOSBIA1217-1G APOSBIA1217-5G
N-Acetyl-D-alanine
N-Acetyl-D-alanine
Vzorec: C₅H₉NO₃
MW: 131,131 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063133
CAS číslo: 19436-52-3
EINECS: 243-066-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR