Tisk…

Acetohydroxamic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: N-Hydroxyacetamide

BIA1030-5G BIA1030-25G
APOSBIA1030-5GEA 705 CZK
APOSBIA1030-5G APOSBIA1030-25G
Acetohydroxamic acid
Acetohydroxamic acid
Vzorec: C₂H₅NO₂
MW: 75.0666 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009994
CAS číslo: 546-88-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR