Tisk…

2,3,4,6-Tetra-O-acetyl-α-D-galactopyranosyl bromide

Dodavatel: Apollo Scientific
Varování

Synonyms: Acetobromo-α-D-galactose, (2R,3S,4S,5R,6R)-2-(acetoxymethyl)-6-bromotetrahydro-2H-pyran-3,4,5-triyl triacetate, Tetra-O-acetyl-α-D-galactopyranosyl bromide, Acetobromo-a-D-galactose

BIA1026-100G BIA1026-25G BIA1026-50G
APOSBIA1026-100GEA 17990 CZK
APOSBIA1026-100G APOSBIA1026-25G APOSBIA1026-50G
2,3,4,6-Tetra-O-acetyl-α-D-galactopyranosyl bromide
2,3,4,6-Tetra-O-acetyl-alpha-D-galactopyranosyl bromide

Stabilised with 1% CaCO3

Vzorec: C₁₄H₁₉BrO₉
MW: 411,2 g/mol
Bod tání: 83…85 °C
Storage Temperature: Freezer
MDL Number: MFCD00063686
CAS číslo: 3068-32-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR