Tisk…

Amikacin disulphate

Dodavatel: Apollo Scientific
Varování

Synonyms: (S)-4-Amino-N-((1R,2S,3S,4R,5S)-5-amino-2-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4-(((2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-3-hydroxycyclohexyl)-2-hydroxybutanamide bis(sulphate)

BIA1010-25G BIA1010-5G
APOSBIA1010-25GEA 10580 CZK
APOSBIA1010-25G APOSBIA1010-5G
Amikacin disulphate
Amikacin disulphate

Potency 674-786 ug per mg

Vzorec: C₂₂H₄₃N₅O₁₃·2H₂SO₄
MW: 781,77 g/mol
Bod tání: 220…230 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00167475
CAS číslo: 39831-55-5
EINECS: 254-658-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR