Tisk…

Adenine sulphate ≥99%

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Aminopurine hemisulphate salt, 7H-Purin-6-amine hemisulphate, Adenine hemisulphate salt

BIA0543-100G BIA0543-25G BIA0543-500G
APOSBIA0543-100GEA 3110 CZK
APOSBIA0543-100G APOSBIA0543-25G APOSBIA0543-500G
Adenine sulphate ≥99%
Adenine sulphate

Cytokin growth regulatorPlant tissue culture tested.

Vzorec: C₅H₅N₅·½H₂SO₄
Bod tání: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00213655
CAS číslo: 321-30-2
EINECS: 206-286-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR