Tisk…

Alizarin red S (sodium salt)

Dodavatel: Apollo Scientific
Varování

BIA0326-100G BIA0326-25G
APOSBIA0326-100GEA 2070 CZK
APOSBIA0326-100G APOSBIA0326-25G
Alizarin red S (sodium salt)
Alizarin red S (sodium salt)

C.I. 58005

Vzorec: C₁₄H₇NaO₇S
Storage Temperature: Ambient
MDL Number: MFCD00013049
CAS číslo: 130-22-3
EINECS: 204-981-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR