Tisk…

(S)-Methyl-2-aminopropanoate hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: H-Ala-OMe·HCl, H-Ala-OMe.HCl, L-Alanine methyl ester hydrochloride, (S)-2-Aminopropionic Acid Methyl Ester Hydrochloride

BIA0121-25G BIA0121-5G
APOSBIA0121-25GEA 1080 CZK
APOSBIA0121-25G APOSBIA0121-5G
(S)-Methyl-2-aminopropanoate hydrochloride
(S)-Methyl-2-aminopropanoate hydrochloride
Vzorec: C₄H₉NO₂·HCl
MW: 139,6 g/mol
Bod tání: 109…111 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063663
CAS číslo: 2491-20-5
EINECS: 219-652-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR