Alloxan monohydrate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Pyrimidine-2,4,5,6(1H,3H)-tetraone hydrate
BIA0118-100G
BIA0118-250G
BIA0118-50G
APOSBIA0118-100GEA
6050
CZK
APOSBIA0118-100G
APOSBIA0118-250G
APOSBIA0118-50G
Alloxan monohydrate
Alloxan monohydrate
Vzorec:
C₄H₄N₂O₅ MW: 160,09 g/mol Bod tání: 255 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006031 CAS číslo: 2244-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...